Anti-SLC11A2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-3577R-A750
BOSSBS-3577R-A750EA 899 EUR
BOSSBS-3577R-A750
Anti-SLC11A2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Important in metal transport, in particular iron. Can also transport manganese, cobalt, cadmium, nickel, vanadium and lead. Involved in apical iron uptake into duodenal enterocytes. Involved in iron transport from acidified endosomes into the cytoplasm of erythroid precursor cells. May play an important role in hepatic iron accumulation and tissue iron distribution. May serve to import iron into the mitochondria.

Type: Primary
Antigen: SLC11A2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 251-350/568
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat; Chicken
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR