Anti-SLC11A2 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-3577R-CY5.5
BOSSBS-3577R-CY5.5EA 909 EUR
BOSSBS-3577R-CY5.5
Anti-SLC11A2 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Important in metal transport, in particular iron. Can also transport manganese, cobalt, cadmium, nickel, vanadium and lead. Involved in apical iron uptake into duodenal enterocytes. Involved in iron transport from acidified endosomes into the cytoplasm of erythroid precursor cells. May play an important role in hepatic iron accumulation and tissue iron distribution. May serve to import iron into the mitochondria.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLC11A2
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR