Anti-SLC11A2 Rabbit Polyclonal Antibody (Cy7®)
Lieferant: Bioss
BS-3577R-CY7
BOSSBS-3577R-CY7EA 901 EUR
BOSSBS-3577R-CY7
Anti-SLC11A2 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Important in metal transport, in particular iron. Can also transport manganese, cobalt, cadmium, nickel, vanadium and lead. Involved in apical iron uptake into duodenal enterocytes. Involved in iron transport from acidified endosomes into the cytoplasm of erythroid precursor cells. May play an important role in hepatic iron accumulation and tissue iron distribution. May serve to import iron into the mitochondria.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SLC11A2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat