Anti-PPAR alpha Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-3614R-A680
BOSSBS-3614R-A680EA 890 EUR
BOSSBS-3614R-A680
Anti-PPAR alpha Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Ligand-activated transcription factor. Key regulator of lipid metabolism. Activated by the endogenous ligand 1-palmitoyl-2-oleoyl-sn-glycerol-3-phosphocholine (16:0/18:1-GPC). Activated by oleylethanolamide, a naturally occurring lipid that regulates satiety. Receptor for peroxisome proliferators such as hypolipidemic drugs and fatty acids. Regulates the peroxisomal beta-oxidation pathway of fatty acids. Functions as transcription activator for the ACOX1 and P450 genes. Transactivation activity requires heterodimerization with RXRA and is antagonized by NR2C2. May be required for the propagation of clock information to metabolic pathways regulated by PER2.
Type: Primary
Antigen: PPAR alpha
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 301-400/468
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat; Chicken; Others