Anti-NCF/NCF4/p40phox Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-3616R-A555
BOSSBS-3616R-A555EA 893 EUR
BOSSBS-3616R-A555
Anti-NCF/NCF4/p40phox Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
NCF4 is a cytosolic oxidase associated protein. It is a component of the nicotinamide adenine dinucleotide phosphate oxidase, which mediates down regulation of NADPH oxidase through interactions with its SH3 domain. NCF4 is associated with p67 phox but is absent in patients with chronic granulomatous disease who lack p67 phox.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NCF/NCF4/p40phox
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR