Anti-NCF/NCF4/p40phox Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Lieferant: Bioss
BS-3616R-A647
BOSSBS-3616R-A647EA 893 EUR
BOSSBS-3616R-A647
Anti-NCF/NCF4/p40phox Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
NCF4 is a cytosolic oxidase associated protein. It is a component of the nicotinamide adenine dinucleotide phosphate oxidase, which mediates down regulation of NADPH oxidase through interactions with its SH3 domain. NCF4 is associated with p67 phox but is absent in patients with chronic granulomatous disease who lack p67 phox.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: NCF/NCF4/p40phox
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat