Anti-NCF/NCF4/p40phox Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-3616R-A750
BOSSBS-3616R-A750EA 890 EUR
BOSSBS-3616R-A750
Anti-NCF/NCF4/p40phox Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
NCF4 is a cytosolic oxidase associated protein. It is a component of the nicotinamide adenine dinucleotide phosphate oxidase, which mediates down regulation of NADPH oxidase through interactions with its SH3 domain. NCF4 is associated with p67 phox but is absent in patients with chronic granulomatous disease who lack p67 phox.

Type: Primary
Antigen: NCF/NCF4/p40phox
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 121-220/339
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR