Anti-HSD11B2 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-3618R-CY3
BOSSBS-3618R-CY3EA 901 EUR
BOSSBS-3618R-CY3
Anti-HSD11B2 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Catalyzes the conversion of cortisol to the inactive metabolite cortisone. Modulates intracellular glucocorticoid levels, thus protecting the nonselective mineralocorticoid receptor from occupation by glucocorticoids.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HSD11B2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR