Anti-PPP1R8 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-3658R-CY5.5
BOSSBS-3658R-CY5.5EA 900 EUR
BOSSBS-3658R-CY5.5
Anti-PPP1R8 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Inhibitor subunit of the major nuclear protein phosphatase 1 (PP1). It has RNA binding activity but does not cleave RNA and may target PP1 to RNA associated substrates. May also be involved in pre mRNA splicing. Binds DNA and might act as a transcriptional repressor. Seems to be required for cell proliferation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PPP1R8
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR