Anti-DACH2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-3659R-A488
BOSSBS-3659R-A488EA 893 EUR
BOSSBS-3659R-A488
Anti-DACH2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
DACH2 is a transcription factor that is involved in regulation of organogenesis. It seems to be a regulator for SIX1 and SIX6 and may act as a corepressor of SIX6 in regulating proliferation by directly repressing cyclin-dependent kinase inhibitors, including the p27Kip1 promoter. It probably binds to DNA via its DACH box-N domain.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DACH2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR