Anti-RALDH2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-3676R-A680
BOSSBS-3676R-A680EA 874 EUR
BOSSBS-3676R-A680
Anti-RALDH2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Recognises as substrates free retinal and cellular retinol-binding protein-bound retinal. Does metabolize octanal and decanal but does not metabolize citral, benzaldehyde, acetaldehyde and propanal efficiently (By similarity).

Type: Primary
Antigen: RALDH2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 441-518/518
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat; Zebrafish
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR