Anti-APC Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Lieferant: Bioss
BS-3680R-A350
BOSSBS-3680R-A350EA 893 EUR
BOSSBS-3680R-A350
Anti-APC Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Tumor suppressor. Promotes rapid degradation of CTNNB1 and participates in Wnt signaling as a negative regulator. APC activity is correlated with its phosphorylation state. Activates the GEF activity of SPATA13 and ARHGEF4. Plays a role in hepatocyte growth factor (HGF)-induced cell migration. Required for MMP9 up-regulation via the JNK signaling pathway in colorectal tumor cells. Acts as a mediator of ERBB2-dependent stabilization of microtubules at the cell cortex. It is required for the localization of MACF1 to the cell membrane and this localization of MACF1 is critical for its function in microtubule stabilization.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: APC
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat