Anti-PKC gamma Thr514 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-3729R-A680
BOSSBS-3729R-A680EA 917 EUR
BOSSBS-3729R-A680
Anti-PKC gamma Thr514 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Protein kinase C (PKC) is a family of serine- and threonine-specific protein kinases that can be activated by calcium and second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play distinct roles in cells. The protein encoded by this gene is one of the PKC family members. This protein kinase is expressed solely inMore Product Information