Anti-SCD Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Lieferant: Bioss
BS-3787R-A647
BOSSBS-3787R-A647EA 893 EUR
BOSSBS-3787R-A647
Anti-SCD Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Terminal component of the liver microsomal stearyl-CoA desaturase system, that utilizes O(2) and electrons from reduced cytochrome b5 to catalyze the insertion of a double bond into a spectrum of fatty acyl-CoA substrates including palmitoyl-CoA and stearoyl-CoA.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SCD
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat