Anti-ZNF211 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss
BS-3800R-FITC
BOSSBS-3800R-FITCEA 883 EUR
BOSSBS-3800R-FITC
Anti-ZNF211 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Immunoglobulins belong to a group of related glyco proteins which make up 20% of serum proteins. Antigens and immunoglobulins react to confer immunity to individuals. Immunoglobulins have similar structures of two identical heavy chains and two identical light chains. Both the heavy chains and the light chains are divided into constant and variable regions. The constant regions have the same amino acid sequences between all the immunoglobulin classes. The variable regions have approximately 110 amino acids with high sequence variability. The amino acid sequence of the heavy chain determinesMore Product Information
Bestellen

TECHNISCHE DATEN

Antigen-Symbol ZNF211
Antigen-Name Zinc Finger Protein 211
Environmentally Preferable
Antigen-Synonyme CH2H2-25|ZNFC25|C2H2-25|ZNF-25
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation FITC (Fluorescein Isothiocyanate)
Reaktivität Human
Wirt Rabbit
Gen-ID 10520
Isotyp IgG
ImmunoChemie Yes
Form liquid
Reinigung Purified by Protein A
Konzentration 1 μg/μl
Kreuzadsorption No
Lagerungspuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagertemperatur Store at 4°C for 12 months
Transporttemperatur 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR