Anti-Lambda Light Chain Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-3801R-A750
BOSSBS-3801R-A750EA 890 EUR
BOSSBS-3801R-A750
Anti-Lambda Light Chain Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
All five immunoglobulin classes share the same basic four polypeptide chain structure of two heavy-chains and two light chains. There are five heavy chain types, and two light-chain types (Kappa and Lambda) both having a molecular weight of 22.5kDa. Any heavy-chain type can associate with either light-chain type, but on any immunoglobulin molecule both light-chains are of the same type. Kappa and Lambda consist of a variable region and a constant region and can easily be differentiated by the antigenic properties of the constant region.

Type: Primary
Antigen: Lambda Light Chain
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 41-106/106
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR