Anti-TP Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-3809R-A555
BOSSBS-3809R-A555EA 893 EUR
BOSSBS-3809R-A555
Anti-TP Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
In the presence of inorganic orthophosphate, the platelet-derived endothelial growth factor (PD-ECGF) thymidine phosphorylase (TP) (gliostatin) catalyses the reversible phospholytic cleavage of thymidine and deoxyuridine to their corresponding bases and 2-deoxyribose-1-phosphate. It is both chemotactic and mitogenic for endothelial cells and a non-heparin binding angiogenic factor present in platelets. It is also involved in transformation of fluoropyrimidines, cytotoxic agents used in the treatment of a variety of malignancies, into active cytotoxic metabolites.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TP
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR