Anti-TP/thymidine phosphorylase Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-3809R-A680
BOSSBS-3809R-A680EA 890 EUR
BOSSBS-3809R-A680
Anti-TP/thymidine phosphorylase Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
In the presence of inorganic orthophosphate, the platelet-derived endothelial growth factor (PD-ECGF) thymidine phosphorylase (TP) (gliostatin) catalyses the reversible phospholytic cleavage of thymidine and deoxyuridine to their corresponding bases and 2-deoxyribose-1-phosphate. It is both chemotactic and mitogenic for endothelial cells and a non-heparin binding angiogenic factor present in platelets. It is also involved in transformation of fluoropyrimidines, cytotoxic agents used in the treatment of a variety of malignancies, into active cytotoxic metabolites.
Type: Primary
Antigen: TP/More Product Information