Anti-CCNG2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Lieferant: Bioss
BS-3822R-A488
BOSSBS-3822R-A488EA 901 EUR
BOSSBS-3822R-A488
Anti-CCNG2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Cyclin G2 may play a role in growth regulation and in negative regulation of cell cycle progesssion. Cyclin G2 expression levels increase through the cell cycle to peak in the mid/late-S phase and decrease during G2/M phase. It may also contribute in maintaining the quiescent state of differentiated cells. Cyclin G2 is similar to cyclin A in the cyclin box, although no kinase activity is detected.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CCNG2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat