Anti-CCNG2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss
BS-3822R-FITC
BOSSBS-3822R-FITCEA 909 EUR
BOSSBS-3822R-FITC
Anti-CCNG2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Cyclin G2 may play a role in growth regulation and in negative regulation of cell cycle progesssion. Cyclin G2 expression levels increase through the cell cycle to peak in the mid/late-S phase and decrease during G2/M phase. It may also contribute in maintaining the quiescent state of differentiated cells. Cyclin G2 is similar to cyclin A in the cyclin box, although no kinase activity is detected.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CCNG2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR