Anti-AMBRA1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-3830R-A750
BOSSBS-3830R-A750EA 899 EUR
BOSSBS-3830R-A750
Anti-AMBRA1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Regulates autophagy and development of the nervous system. Involved in autophagy in controlling protein turnover during neuronal development, and in regulating normal cell survival and proliferation (By similarity).

Type: Primary
Antigen: AMBRA1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 221-320/1298
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR