Anti-RHEB Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Lieferant: Bioss
BS-3832R-HRP
BOSSBS-3832R-HRPEA 900 EUR
BOSSBS-3832R-HRP
Anti-RHEB Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Stimulates the phosphorylation of S6K1 and EIF4EBP1 through activation of mTORC1 signaling. Activates the protein kinase activity of mTORC1. Has low intrinsic GTPase activity.
Recommended Dilutions: IHC-P: 1:100-500
Type: Primary
Antigen: RHEB
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat