Anti-CHRDL2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-3860R-A488
BOSSBS-3860R-A488EA 893 EUR
BOSSBS-3860R-A488
Anti-CHRDL2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
CHRDL2 is a novel chordin like protein that can act as a BMP antagonist. Amember of the chordin family of proteins, it contains a signal peptide andthree CR (cysteine-rich repeat) domains. When expressed as a recombinantprotein it is secreted and binds to activin A, but not to BMP-2, -4, -6. Differential expression has been detected in developing chondrocytes, myoblasts, osteoblasts, and osteoarthritic joints. Complex alternative splicing of CHRDL2 potentially results in distinct isoforms that differ at their C termini, in the expression of signal peptide, and in the content of CR domains. CHRDL2 was originally characterized as a novel protein exclusively expressed in breast, lung, and colon tumors.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CHRDL2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR