Anti-GPX3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-3883R-A750
BOSSBS-3883R-A750EA 1350 EUR
BOSSBS-3883R-A750
Anti-GPX3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Protects cells and enzymes from oxidative damage, by catalyzing the reduction of hydrogen peroxide, lipid peroxides and organic hydroperoxide, by glutathione.

Type: Primary
Antigen: GPX3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-200/226
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR