Anti-GPX3 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-3883R-CY5
BOSSBS-3883R-CY5EA 1350 EUR
BOSSBS-3883R-CY5
Anti-GPX3 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Protects cells and enzymes from oxidative damage, by catalyzing the reduction of hydrogen peroxide, lipid peroxides and organic hydroperoxide, by glutathione.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GPX3
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR