Anti-NDUFS3 Rabbit Polyclonal Antibody (Cy7®)
Lieferant: Bioss
BS-3888R-CY7
BOSSBS-3888R-CY7EA 900 EUR
BOSSBS-3888R-CY7
Anti-NDUFS3 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Core subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I) that is believed to belong to the minimal assembly required for catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone (By similarity).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: NDUFS3
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat