Anti-cGMP Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-3892R-CY5.5
BOSSBS-3892R-CY5.5EA 900 EUR
BOSSBS-3892R-CY5.5
Anti-cGMP Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Cyclic guanosine monophosphate (cGMP) serves as a second messenger in a manner similar to that observed with cAMP. Peptide hormones, such as the natriuretic factors, activate receptors that are associated with membrane-bound guanylate cyclase (GC). Receptor activation of GC leads to the conversion of GTP to cGMP. Nitric oxide (NO) also stimulates cGMP production by activating soluble GC, perhaps by binding to the heme moiety of the enzyme. Similar to cAMP, cGMP mediates most of its intracellular effects through the activation of specific cGMP dependent protein kinases (PKG).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: cGMP
Clonality: Polyclonal
Clone: 0
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR