Anti-SOD4/CCS Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-3896R-A750
BOSSBS-3896R-A750EA 890 EUR
BOSSBS-3896R-A750
Anti-SOD4/CCS Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Superoxide dismutase (SOD) is an antioxidant enzyme involved in the defence system against reactive oxygen species (ROS). SOD catalyses the dismutation reaction of superoxide radical anion (O2-) to hydrogen peroxide, which is then catalysed to innocuous O2 and H2O by glutathione peroxidase and catalase. Several classes of SOD have been identified. These include intracellular copper, zinc SOD (Cu, Zn-SOD/SOD-1), mitochondrial manganese SOD (Mn-SOD/SOD-2) and extracellular Cu, Zn-SOD (EC-SOD/SOD-3). SOD1 is found in all eukaryotic species as a homodimeric 32 kDa enzyme containing one each of Cu and Zn ion per subunit. The manganese containing 80 kDa tetrameric enzyme SOD2, is located in the mitochondrial matrix in close proximity to a primary endogenous source of superoxide, the mitochondrial respiratory chain. SOD3 is a heparin-binding multimer of disulfide-linked dimers, primarily expressed in human lungs, vessel walls and airways. SOD4 is a copper chaperone for superoxide dismutase (CCS), which specifically delivers Cu to copper/zinc superoxide dismutase. CCS may activate copper/zinc superoxide dismutase through direct insertion of the Cu cofactor.

Type: Primary
Antigen: SOD4/CCS
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 201-274/274
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR