Anti-CYP24A1 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-3899R-CY5
BOSSBS-3899R-CY5EA 900 EUR
BOSSBS-3899R-CY5
Anti-CYP24A1 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Has a role in maintaining calcium homeostasis. Catalyzes the NADPH-dependent 24-hydroxylation of calcidiol (25-hydroxyvitamin D(3)) and calcitriol (1-alpha,25-dihydroxyvitamin D(3)). The enzyme can perform up to 6 rounds of hydroxylation of calcitriol leading to calcitroic acid. It also shows 23-hydroxylating activity leading to 1-alpha,25-dihydroxyvitamin D(3)-26,23-lactone as end product.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CYP24A1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR