Anti-CYP2R1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-3900R-A555
BOSSBS-3900R-A555EA 893 EUR
BOSSBS-3900R-A555
Anti-CYP2R1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
CYP2R1 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are mono-oxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This enzyme is a microsomal vitamin D hydroxylase that converts vitamin D into the active ligand for the vitamin D receptor.Defects in CYP2R1 are a cause of 25-hydroxyvitamin D(3) deficiency, also known as pseudovitamin D(3) deficiency rickets due to 25-hydroxylase deficiency. First described in patients who had rickets at a young age despite a history of adequate vitamin D intake. The patients sera had low calcium concentrations, low phosphate concentrations, elevated alkaline phosphatase activity and low levels of 25-hydroxyvitamin D.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CYP2R1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR