Anti-PNMT Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss
BS-3912R-A350
BOSSBS-3912R-A350EA 876 EUR
BOSSBS-3912R-A350
Anti-PNMT Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Converts noradrenaline to adrenaline.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PNMT
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR