Anti-PNMT Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Lieferant: Bioss
BS-3912R-A555
BOSSBS-3912R-A555EA 876 EUR
BOSSBS-3912R-A555
Anti-PNMT Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Converts noradrenaline to adrenaline.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PNMT
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat