Anti-PNMT Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-3912R-A750
BOSSBS-3912R-A750EA 874 EUR
BOSSBS-3912R-A750
Anti-PNMT Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Converts noradrenaline to adrenaline.

Type: Primary
Antigen: PNMT
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-200/282
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR