Anti-PNMT Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss
BS-3912R-HRP
BOSSBS-3912R-HRPEA 883 EUR
BOSSBS-3912R-HRP
Anti-PNMT Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Converts noradrenaline to adrenaline.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: PNMT
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR