Anti-ACLY Rabbit Polyclonal Antibody (Cy3®)
Lieferant: Bioss
BS-3929R-CY3
BOSSBS-3929R-CY3EA 900 EUR
BOSSBS-3929R-CY3
Anti-ACLY Rabbit Polyclonal Antibody (Cy3®)
Antikörper
ATP citrate lyase is the primary enzyme responsible for the synthesis of cytosolic acetyl-CoA in many tissues. The enzyme is a tetramer (relative molecular weight approximately 440,000) of apparently identical subunits. It catalyzes the formation of acetyl-CoA and oxaloacetate from citrate and CoA with a concomitant hydrolysis of ATP to ADP and phosphate. The product, acetyl-CoA, serves several important biosynthetic pathways, including lipogenesis and cholesterogenesis. In nervous tissue, ATP citrate-lyase may be involved in the biosynthesis of acetylcholine. Two transcript variantsMore Product Information