Anti-FH Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-3938R
BOSSBS-3938REA 725 EUR
BOSSBS-3938R
Anti-FH Rabbit Polyclonal Antibody
Antikörper
Defects in FH are the cause of fumarase deficiency (FHD)also known as fumaricaciduria. FHD is characterized by progressive encephalopathy, developmental delay, hypotonia, cerebral atrophy and lactic and pyruvic acidemia.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: Fumarate hydratase/FH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR