Anti-FH Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Lieferant: Bioss
BS-3938R-A488
BOSSBS-3938R-A488EA 893 EUR
BOSSBS-3938R-A488
Anti-FH Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Defects in FH are the cause of fumarase deficiency (FHD)also known as fumaricaciduria. FHD is characterized by progressive encephalopathy, developmental delay, hypotonia, cerebral atrophy and lactic and pyruvic acidemia.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Fumarate hydratase/FH
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat