Anti-FH Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-3938R-A555
BOSSBS-3938R-A555EA 893 EUR
BOSSBS-3938R-A555
Anti-FH Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Defects in FH are the cause of fumarase deficiency (FHD)also known as fumaricaciduria. FHD is characterized by progressive encephalopathy, developmental delay, hypotonia, cerebral atrophy and lactic and pyruvic acidemia.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Fumarate hydratase/FH
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR