Anti-Fumarate hydratase/FH Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-3938R-A680
BOSSBS-3938R-A680EA 890 EUR
BOSSBS-3938R-A680
Anti-Fumarate hydratase/FH Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Defects in FH are the cause of fumarase deficiency (FHD)also known as fumaricaciduria. FHD is characterised by progressive encephalopathy, developmental delay, hypotonia, cerebral atrophy and lactic and pyruvic acidemia.

Type: Primary
Antigen: Fumarate hydratase/FH
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 411-510/510
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR