Anti-FH Rabbit Polyclonal Antibody (Cy5®)
Lieferant: Bioss
BS-3938R-CY5
BOSSBS-3938R-CY5EA 900 EUR
BOSSBS-3938R-CY5
Anti-FH Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Defects in FH are the cause of fumarase deficiency (FHD)also known as fumaricaciduria. FHD is characterized by progressive encephalopathy, developmental delay, hypotonia, cerebral atrophy and lactic and pyruvic acidemia.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Fumarate hydratase/FH
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat