Anti-FH Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-3938R-CY7
BOSSBS-3938R-CY7EA 900 EUR
BOSSBS-3938R-CY7
Anti-FH Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Defects in FH are the cause of fumarase deficiency (FHD)also known as fumaricaciduria. FHD is characterized by progressive encephalopathy, developmental delay, hypotonia, cerebral atrophy and lactic and pyruvic acidemia.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Fumarate hydratase/FH
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR