Anti-FH Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss
BS-3938R-HRP
BOSSBS-3938R-HRPEA 900 EUR
BOSSBS-3938R-HRP
Anti-FH Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Defects in FH are the cause of fumarase deficiency (FHD)also known as fumaricaciduria. FHD is characterized by progressive encephalopathy, developmental delay, hypotonia, cerebral atrophy and lactic and pyruvic acidemia.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: Fumarate hydratase/FH
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR