Anti-GNG2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Lieferant: Bioss
BS-3944R-A350
BOSSBS-3944R-A350EA 893 EUR
BOSSBS-3944R-A350
Anti-GNG2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
GNG2 (Guanine nucleotide binding protein (G protein), gamma 2) is a member of the guanine nucleotide-binding proteins (G proteins) family. Heterotrimeric G proteins are involved as a modulator or transducer in various transmembrane signaling systems. The specificity of a G protein-receptor interaction is primarily mediated by the gamma subunit.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GNG2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat