Anti-GNG2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-3944R-A680
BOSSBS-3944R-A680EA 890 EUR
BOSSBS-3944R-A680
Anti-GNG2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
GNG2 (Guanine nucleotide binding protein (G protein), gamma 2) is a member of the guanine nucleotide-binding proteins (G proteins) family. Heterotrimeric G proteins are involved as a modulator or transducer in various transmembrane signaling systems. The specificity of a G protein-receptor interaction is primarily mediated by the gamma subunit.

Type: Primary
Antigen: GNG2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 2-50/71
Host: Rabbit
Isotype: IgG
Reactivity: Zebrafish
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR