Anti-IDH3A Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-3946R-A680
BOSSBS-3946R-A680EA 899 EUR
BOSSBS-3946R-A680
Anti-IDH3A Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Isocitrate dehydrogenases catalyse the oxidative decarboxylation of isocitrate to 2-oxoglutarate. These enzymes belong to two distinct subclasses, one of which utilizes NAD(+) as the electron acceptor and the other NADP(+). Five isocitrate dehydrogenases have been reported: three NAD(+)-dependent isocitrate dehydrogenases, which localise to the mitochondrial matrix, and two NADP(+)-dependent isocitrate dehydrogenases, one of which is mitochondrial and the other predominantly cytosolic. NAD(+)-dependent isocitrate dehydrogenases catalyse the allosterically regulated rate-limiting step of the tricarboxylic acid cycle. Each isozyme is a heterotetramer that is composed of two alpha subunits, one beta subunit, and one gamma subunit. IDH3A is the alpha subunit of one isozyme of NAD(+)-dependent isocitrate dehydrogenase.

Type: Primary
Antigen: IDH3A
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 281-366/366
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat; Others
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR