Anti-COX1/MTCO1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-3953R-A750
BOSSBS-3953R-A750EA 899 EUR
BOSSBS-3953R-A750
Anti-COX1/MTCO1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Cytochrome c oxidase is the component of the respiratory chain that catalyses the reduction of oxygen to water. Subunits 1-3 form the functional core of the enzyme complex. CO I is the catalytic subunit of the enzyme. Electrons originating in cytochrome c are transferred via the copper A centre of subunit 2 and heme A of subunit 1 to the bimetallic centre formed by heme A3 and copper B. This protein is involved in the pathway oxidative phosphorylation, which is part of Energy metabolism.

Type: Primary
Antigen: COX1/MTCO1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 401-500/513
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR