Anti-COX1 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-3953R-CY5.5
BOSSBS-3953R-CY5.5EA 909 EUR
BOSSBS-3953R-CY5.5
Anti-COX1 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Cytochrome c oxidase subunit I (COI or MTCO1) is one of three mitochondrial DNA (mtDNA) encoded subunits (MTCO1, MTCO2, MTCO3) of respiratory Complex IV. Complex IV is located within the mitochondrial inner membrane and is the third and final enzyme of the electron transport chain of mitochondrial oxidative phosphorylation. Complex IV is composed of 13 polypeptides. Subunits I, II, and III (MTCO1, MTCO2, MTCO3) are encoded by mtDNA while subunits IV, Va, Vb, VIa, VIb, VIc, VIIa, VIIb, VIIc, and VIII are nuclear encoded. Mammalian MTCO1 has 12 membrane-spanning alpha-helices (I to XII).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: COX1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR