Anti-NDUFA10 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-3957R-A750
BOSSBS-3957R-A750EA 890 EUR
BOSSBS-3957R-A750
Anti-NDUFA10 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
Type: Primary
Antigen: NDUFA10
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 201-300/355
Host: Rabbit
Isotype: IgG
Reactivity: Mouse