Anti-NDUFA8 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Lieferant: Bioss
BS-3958R-A647
BOSSBS-3958R-A647EA 893 EUR
BOSSBS-3958R-A647
Anti-NDUFA8 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: NDUFA8
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat