Anti-NDUFV1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-3959R-A750
BOSSBS-3959R-A750EA 890 EUR
BOSSBS-3959R-A750
Anti-NDUFV1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Core subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I) that is believed to belong to the minimal assembly required for catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone (By similarity).

Type: Primary
Antigen: NDUFV1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 41-140/464
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat; Others
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR